2-Acetyl-3,5-dimethylpyrazine 95%
Dodavatel: Apollo Scientific
Synonyms:
1-(3,5-Dimethylpyrazin-2-yl)ethanone
OR904583-25G
OR904583-5G
APOSOR904583-25GEA
6550
CZK
APOSOR904583-25G
APOSOR904583-5G
2-Acetyl-3,5-dimethylpyrazine 95%
2-Acetyl-3,5-dimethylpyrazine
Vzorec:
C₈H₁₀N₂O MW: 150.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055023 CAS číslo: 54300-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...