Tisk…

1-Pyridin-3-ylmethyl-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR904526-1G OR904526-250MG
APOSOR904526-1GEA 19440 CZK
APOSOR904526-1G APOSOR904526-250MG
1-Pyridin-3-ylmethyl-1H-pyrrole-2-carbaldehyde 95%
1-Pyridin-3-ylmethyl-1H-pyrrole-2-carbaldehyde
Vzorec: C₁₁H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD02665151
CAS číslo: 186801-36-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR