Tisk…

Ethyl-2-oxovalerate 96%

Dodavatel: Apollo Scientific

OR904458-100G OR904458-25G
APOSOR904458-100GEA 33400 CZK
APOSOR904458-100G APOSOR904458-25G
Ethyl-2-oxovalerate 96%
Ethyl-2-oxovalerate
Vzorec: C₇H₁₂O₃
MW: 144,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410882
CAS číslo: 50461-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR