4-Ethyl-2-methylthiazole-5-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR904449-1G
OR904449-250MG
APOSOR904449-1GEA
15310
CZK
APOSOR904449-1G
APOSOR904449-250MG
4-Ethyl-2-methylthiazole-5-carboxylic acid 95%
4-Ethyl-2-methylthiazole-5-carboxylic acid
Vzorec:
C₇H₉NO₂S MW: 171,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12027007 CAS číslo: 119778-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...