Tisk…

3,3-Dimethylpiperidin-4-ol 95%

Dodavatel: Apollo Scientific

OR904308-1G OR904308-250MG
APOSOR904308-1GEA 4770 CZK
APOSOR904308-1G APOSOR904308-250MG
3,3-Dimethylpiperidin-4-ol 95%
3,3-Dimethylpiperidin-4-ol
Vzorec: C7H15NO
MW: 129.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044222
CAS číslo: 373603-88-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR