Tisk…

1-Ethyl-3-methyl-1H-pyrazole-4-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR904228-1G OR904228-250MG
APOSOR904228-1GEA 10350 CZK
APOSOR904228-1G APOSOR904228-250MG
1-Ethyl-3-methyl-1H-pyrazole-4-carbaldehyde 95%
1-Ethyl-3-methyl-1H-pyrazole-4-carbaldehyde
Vzorec: C₇H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD02056541
CAS číslo: 676348-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR