Tisk…

Thiophene-2-carboxaldoxime 95%

Dodavatel: Apollo Scientific

OR904201-25G
APOSOR904201-25GEA 2400 CZK
APOSOR904201-25G
Thiophene-2-carboxaldoxime 95%
Thiophene-2-carboxaldoxime
Vzorec: C₅H₅NOS
MW: 127,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022509
CAS číslo: 29683-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR