2-Methoxynicotinic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid
OR9042-5G
OR9042-1G
APOSOR9042-5GEA
500
CZK
APOSOR9042-5G
APOSOR9042-1G
2-Methoxynicotinic acid 98%
2-Methoxynicotinic acid
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod tání: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085110 CAS číslo: 16498-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...