Tisk…

3-Ethynylpyridine 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Pyridylacetylene

OR904149-25G
APOSOR904149-25GEA 5890 CZK
APOSOR904149-25G
3-Ethynylpyridine 97%
3-Ethynylpyridine
Vzorec: C₇H₅N
MW: 103,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02177459
CAS číslo: 2510-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR