N-Phenylisonicotinamide 98%
Dodavatel: Apollo Scientific
Synonyms:
N-Phenyl isonicotinicamide, N-Phenyl-4-pyridinecarboxamide
OR904044-5G
APOSOR904044-5GEA
1700
CZK
APOSOR904044-5G
N-Phenylisonicotinamide 98%
N-Phenylisonicotinamide
Vzorec:
C₁₂H₁₀N₂O MW: 198,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00459246 CAS číslo: 3034-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...