(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl N-(Triethylammoniosulphonyl)carbamate, Burgess Reagent
OR904017-1G
OR904017-5G
OR904017-25G
APOSOR904017-1GEA
953
CZK
APOSOR904017-1G
APOSOR904017-5G
APOSOR904017-25G
(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt 95%
(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt
Vzorec:
C₈H₁₈N₂O₄S MW: 238.30 g/mol Bod tání: 76…79 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077815 CAS číslo: 29684-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...