3-Ethyl-1H-pyrazole-5-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR904005-100MG
OR904005-1G
OR904005-250MG
APOSOR904005-100MGEA
5390
CZK
APOSOR904005-100MG
APOSOR904005-1G
APOSOR904005-250MG
3-Ethyl-1H-pyrazole-5-carboxylic acid 95%
3-Ethyl-1H-pyrazole-5-carboxylic acid
Vzorec:
C₆H₈N₂O₂ MW: 140,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06742804 CAS číslo: 4027-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...