Tisk…

4-(Thiazol-2-yloxy)phenylamine 96%

Dodavatel: Apollo Scientific

OR903971-1G OR903971-5G
APOSOR903971-1GEA 4040 CZK
APOSOR903971-1G APOSOR903971-5G
4-(Thiazol-2-yloxy)phenylamine 96%
4-(Thiazol-2-yloxy)phenylamine
Vzorec: C₉H₈N₂OS
Storage Temperature: Ambient
MDL Number: MFCD09743704
CAS číslo: 105350-49-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR