Tisk…

1-Methyl-5-phenyl-1H-imidazol-2-ylamine 95%

Dodavatel: Apollo Scientific

OR903941-1G OR903941-250MG
APOSOR903941-1GEA 24600 CZK
APOSOR903941-1G APOSOR903941-250MG
1-Methyl-5-phenyl-1H-imidazol-2-ylamine 95%
1-Methyl-5-phenyl-1H-imidazol-2-ylamine
Vzorec: C₁₀H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD05668690
CAS číslo: 787586-80-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR