Tisk…

2-Morpholinopyrimidin-4-ol 95%

Dodavatel: Apollo Scientific

OR903939-1G OR903939-250MG
APOSOR903939-1GEA 2030 CZK
APOSOR903939-1G APOSOR903939-250MG
2-Morpholinopyrimidin-4-ol 95%
2-Morpholinopyrimidin-4-ol
Vzorec: C₈H₁₁N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00234443
CAS číslo: 19810-79-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR