6-Bromo-8-chloroimidazo[1,2-a]pyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR903923-1G
OR903923-250MG
APOSOR903923-1GEA
1660
CZK
APOSOR903923-1G
APOSOR903923-250MG
6-Bromo-8-chloroimidazo[1,2-a]pyridine 97%
6-Bromo-8-chloroimidazo[1,2-a]pyridine
Vzorec:
C₇H₄BrClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11518983 CAS číslo: 474708-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...