Tisk…

(1-Ethyl-4-piperidinyl)methanol 95%

Dodavatel: Apollo Scientific

Synonyms: (1-Ethylpiperidin-4-yl)methanol

OR903901-1G OR903901-5G
APOSOR903901-1GEA 1540 CZK
APOSOR903901-1G APOSOR903901-5G
(1-Ethyl-4-piperidinyl)methanol 95%
(1-Ethyl-4-piperidinyl)methanol
Vzorec: C₈H₁₇NO
MW: 143,2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059823
CAS číslo: 90226-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR