4,5-Dichloropyrimidine 98%
Dodavatel: Apollo Scientific
OR903869-1G
OR903869-5G
APOSOR903869-1GEA
1820
CZK
APOSOR903869-1G
APOSOR903869-5G
4,5-Dichloropyrimidine 98%
4,5-Dichloropyrimidine
Vzorec:
C₄H₂Cl₂N₂ MW: 148,9800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09743880 CAS číslo: 6554-61-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...