2-Chloro-9-methyl-9H-purine 95%
Dodavatel: Apollo Scientific
OR903840-1G
OR903840-250MG
APOSOR903840-1GEA
21490
CZK
APOSOR903840-1G
APOSOR903840-250MG
2-Chloro-9-methyl-9H-purine 95%
2-Chloro-9-methyl-9H-purine
Vzorec:
C₆H₅ClN₄ MW: 168,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07440150 CAS číslo: 2346-73-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...