Tisk…

(S)-2-Hydroxy-4-phenylbutyric acid 95%

Dodavatel: Apollo Scientific

OR903813-10G OR903813-1G OR903813-5G
APOSOR903813-10GEA 4930 CZK
APOSOR903813-10G APOSOR903813-1G APOSOR903813-5G
(S)-2-Hydroxy-4-phenylbutyric acid 95%
(S)-2-Hydroxy-4-phenylbutyric acid
Vzorec: C₁₀H₁₂O₃
MW: 180,2005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656492
CAS číslo: 115016-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR