Tisk…

2-Amino-6-bromopyrazine 98%

Dodavatel: Apollo Scientific

OR903808-1G OR903808-5G
APOSOR903808-1GEA 2440 CZK
APOSOR903808-1G APOSOR903808-5G
2-Amino-6-bromopyrazine 98%
2-Amino-6-bromopyrazine
Vzorec: C₄H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD10697806
CAS číslo: 54237-53-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR