Tisk…

N-Butyl-2-chloroacetamide 95%

Dodavatel: Apollo Scientific

OR903763-1G OR903763-5G
APOSOR903763-1GEA 1410 CZK
APOSOR903763-1G APOSOR903763-5G
N-Butyl-2-chloroacetamide 95%
N-Butyl-2-chloroacetamide
Vzorec: C₆H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD00776256
CAS číslo: 5349-24-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR