5-Isobutylimidazolidine-2,4-dione 95%
Dodavatel: Apollo Scientific
OR903760-1G
OR903760-250MG
OR903760-5G
APOSOR903760-1GEA
8500
CZK
APOSOR903760-1G
APOSOR903760-250MG
APOSOR903760-5G
5-Isobutylimidazolidine-2,4-dione 95%
5-Isobutylimidazolidine-2,4-dione
Vzorec:
C₇H₁₂N₂O₂ MW: 156,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00238853 CAS číslo: 67337-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...