4-Chlorobenzyl isocyanide 95%
Dodavatel: Apollo Scientific
OR903740-100MG
OR903740-1G
OR903740-250MG
APOSOR903740-100MGEA
4770
CZK
APOSOR903740-100MG
APOSOR903740-1G
APOSOR903740-250MG
4-Chlorobenzyl isocyanide 95%
4-Chlorobenzyl isocyanide
Vzorec:
C₈H₆ClN Storage Temperature: Ambient |
CAS číslo:
39546-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...