Tisk…

3,4-Dichloro-1,2,5-thiadiazole 98%

Dodavatel: Apollo Scientific

OR903574-100G OR903574-25G
APOSOR903574-100GEA 1510 CZK
APOSOR903574-100G APOSOR903574-25G
3,4-Dichloro-1,2,5-thiadiazole 98%
3,4-Dichloro-1,2,5-thiadiazole
Vzorec: C₂Cl₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD00010072
CAS číslo: 5728-20-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR