Tisk…

Imidazo[1,5-a]pyridine-6-carboxylic acid 98%

Dodavatel: Apollo Scientific

OR903526-1G OR903526-250MG
APOSOR903526-1GEA 12900 CZK
APOSOR903526-1G APOSOR903526-250MG
Imidazo[1,5-a]pyridine-6-carboxylic acid 98%
Imidazo[1,5-a]pyridine-6-carboxylic acid
Vzorec: C₈H₆N₂O₂
MW: 162.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042992
CAS číslo: 256935-76-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR