Tisk…

4,8-Dichloroquinazoline 98%

Dodavatel: Apollo Scientific

Synonyms: 4,8-Dichloro-quinazoline

OR903493-1G OR903493-5G
APOSOR903493-1GEA 1950 CZK
APOSOR903493-1G APOSOR903493-5G
4,8-Dichloroquinazoline 98%
4,8-Dichloroquinazoline
Vzorec: C₈H₄Cl₂N₂
MW: 199.400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657224
CAS číslo: 7148-34-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR