Tisk…

3-Chloro-6-(methylsulphonyl)pyridazine 95%

Dodavatel: Apollo Scientific

OR903459-1G OR903459-250MG
APOSOR903459-1GEA 7080 CZK
APOSOR903459-1G APOSOR903459-250MG
3-Chloro-6-(methylsulphonyl)pyridazine 95%
3-Chloro-6-(methylsulphonyl)pyridazine
Vzorec: C5H5ClN2O2S
MW: 192,6234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03931474
CAS číslo: 7145-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR