Tisk…

4-Bromo-6-hydroxypyrimidine 97%

Dodavatel: Apollo Scientific

OR903453-1G OR903453-250MG
APOSOR903453-1GEA 2200 CZK
APOSOR903453-1G APOSOR903453-250MG
4-Bromo-6-hydroxypyrimidine 97%
4-Bromo-6-hydroxypyrimidine
Vzorec: C₄H₃BrN₂O
MW: 174,9834 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223276
CAS číslo: 1086382-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR