5-Bromoimidazo[1,2-a]pyrazine 95%
Dodavatel: Apollo Scientific
OR903328-1G
OR903328-250MG
APOSOR903328-1GEA
4470
CZK
APOSOR903328-1G
APOSOR903328-250MG
5-Bromoimidazo[1,2-a]pyrazine 95%
5-Bromoimidazo[1,2-a]pyrazine
Vzorec:
C₆H₄BrN₃ MW: 198,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08460076 CAS číslo: 87597-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...