Triethyl methanetricarboxylate 98%
Dodavatel: Apollo Scientific
OR903316-100G
OR903316-500G
APOSOR903316-100GEA
3060
CZK
APOSOR903316-100G
APOSOR903316-500G
Triethyl methanetricarboxylate 98%
Triethyl methanetricarboxylate
Vzorec:
C₁₀H₁₆O₆ MW: 232,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009150 CAS číslo: 6279-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...