2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 95%
Dodavatel: Apollo Scientific
OR903289-1G
APOSOR903289-1GEA
2110
CZK
APOSOR903289-1G
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 95%
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile
Vzorec:
C₁₀H₇NO MW: 157,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234685 CAS číslo: 25724-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...