Tisk…

Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 5-chloro-1H-1,2,4-triazole-3-carboxylate

OR903153-1G OR903153-250MG
APOSOR903153-1GEA 2110 CZK
APOSOR903153-1G APOSOR903153-250MG
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate 95%
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate
Vzorec: C₅H₆ClN₃O₂
MW: 175,5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040186
CAS číslo: 774608-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR