2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile 95%
Dodavatel: Apollo Scientific
OR903049-1G
OR903049-250MG
APOSOR903049-1GEA
19440
CZK
APOSOR903049-1G
APOSOR903049-250MG
2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile 95%
2-Amino-4-ethyl-5-methylthiophene-3-carbonitrile
Vzorec:
C₈H₁₀N₂S Storage Temperature: Ambient |
MDL Number:
MFCD01114970 CAS číslo: 4651-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...