Tisk…

2-Phenylthiazole 95%

Dodavatel: Apollo Scientific

OR903003-5G
APOSOR903003-5GEA 2280 CZK
APOSOR903003-5G
2-Phenylthiazole 95%
2-Phenylthiazole
Vzorec: C₉H₇NS
MW: 161,2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272327
CAS číslo: 1826-11-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR