Tisk…

(3-Nitrophenyl)acetonitrile 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Nitrobenzyl cyanide, 2-(3-Nitrophenyl)acetonitrile

OR902997-25G OR902997-5G
APOSOR902997-25GEA 6580 CZK
APOSOR902997-25G APOSOR902997-5G
(3-Nitrophenyl)acetonitrile 97%
(3-Nitrophenyl)acetonitrile
Vzorec: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041249
CAS číslo: 621-50-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR