Tisk…

2-(4-Methylphenyl)pyrrolidine 98%

Dodavatel: Apollo Scientific

OR902962-10G OR902962-1G OR902962-5G
APOSOR902962-10GEA 9330 CZK
APOSOR902962-10G APOSOR902962-1G APOSOR902962-5G
2-(4-Methylphenyl)pyrrolidine 98%
2-(4-Methylphenyl)pyrrolidine
Vzorec: C₁₁H₁₅N
MW: 161,2400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663452
CAS číslo: 62506-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR