tert-Butyl (2-oxopropyl)carbamate 97%
Dodavatel: Apollo Scientific
Synonyms:
(2-Oxopropyl)carbamic acid tert-butyl ester
OR902921-25G
APOSOR902921-25GEA
2400
CZK
APOSOR902921-25G
tert-Butyl (2-oxopropyl)carbamate 97%
tert-Butyl (2-oxopropyl)carbamate
Vzorec:
C₈H₁₅NO₃ MW: 173,2096 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10698678 CAS číslo: 170384-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...