1-Phenyl-1H-pyrrole-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR902874-100MG
OR902874-250MG
OR902874-50MG
APOSOR902874-100MGEA
2410
CZK
APOSOR902874-100MG
APOSOR902874-250MG
APOSOR902874-50MG
1-Phenyl-1H-pyrrole-2-carboxylic acid 95%
1-Phenyl-1H-pyrrole-2-carboxylic acid
Vzorec:
C₁₁H₉NO₂ MW: 187,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05174745 CAS číslo: 78540-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...