5,6-Dichloro-2,3-dicyanopyrazine 96%
Dodavatel: Apollo Scientific
Synonyms:
5,6-Dichloropyrazine-2,3-dicarbonitrile, 5,6-Dichloro-2,3-pyrazinedicarbonitrile
OR902743-25G
APOSOR902743-25GEA
6290
CZK
APOSOR902743-25G
5,6-Dichloro-2,3-dicyanopyrazine 96%
5,6-Dichloro-2,3-dicyanopyrazine
Vzorec:
C₆Cl₂N₄ MW: 198,99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191419 CAS číslo: 56413-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...