Tisk…

N-(tert-Butoxycarbonyl)-L-serine-β-lactone 95%

Dodavatel: Apollo Scientific

OR902727-1G OR902727-250MG
APOSOR902727-1GEA 2610 CZK
APOSOR902727-1G APOSOR902727-250MG
N-(tert-Butoxycarbonyl)-L-serine-β-lactone 95%
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone
Vzorec: C₈H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD01318414
CAS číslo: 98541-64-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR