5,6-Dichloropyrimidin-4-ol 95%
Dodavatel: Apollo Scientific
Synonyms:
5,6-Dichloropyrimidin-4(3H)-one
OR902633-1G
OR902633-250MG
APOSOR902633-1GEA
2820
CZK
APOSOR902633-1G
APOSOR902633-250MG
5,6-Dichloropyrimidin-4-ol 95%
5,6-Dichloropyrimidin-4-ol
Vzorec:
C₄H₂Cl₂N₂O MW: 164,9775 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24393897 CAS číslo: 88982-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...