Tisk…

7-Nitroquinoline 98%

Dodavatel: Apollo Scientific

OR902610-25G OR902610-5G
APOSOR902610-25GEA 8700 CZK
APOSOR902610-25G APOSOR902610-5G
7-Nitroquinoline 98%
7-Nitroquinoline
Vzorec: C₉H₆N₂O₂
MW: 174,1561 g/mol
Bod tání: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00234498
CAS číslo: 613-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR