Cycloheptane-1,3-dione 98%
Dodavatel: Apollo Scientific
Synonyms:
1,3-Cycloheptanedione
OR902471-5G
APOSOR902471-5GEA
3610
CZK
APOSOR902471-5G
Cycloheptane-1,3-dione 98%
Cycloheptane-1,3-dione
Vzorec:
C₇H₁₀O₂ MW: 126,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863735 CAS číslo: 1194-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...