3-Hydroxy-3-phenylpropanoic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
3-Hydroxy-3-phenylpropionic acid
OR902414-25G
APOSOR902414-25GEA
3310
CZK
APOSOR902414-25G
3-Hydroxy-3-phenylpropanoic acid 98%
3-Hydroxy-3-phenylpropanoic acid
Vzorec:
C9H10O3 MW: 166,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00137907 CAS číslo: 3480-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...