Tisk…

1-(5-Chloro-1H-indazol-1-yl)ethanone 95%

Dodavatel: Apollo Scientific

OR902309-1G OR902309-5G
APOSOR902309-1GEA 744 CZK
APOSOR902309-1G APOSOR902309-5G
1-(5-Chloro-1H-indazol-1-yl)ethanone 95%
1-(5-Chloro-1H-indazol-1-yl)ethanone
Vzorec: C₉H₇ClN₂O
MW: 194,6177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109546
CAS číslo: 98083-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR