Tisk…

Benzo[d]oxazol-2-ylmethanol 95%

Dodavatel: Apollo Scientific

OR902294-10G OR902294-1G OR902294-5G
APOSOR902294-10GEA 14020 CZK
APOSOR902294-10G APOSOR902294-1G APOSOR902294-5G
Benzo[d]oxazol-2-ylmethanol 95%
Benzo[d]oxazol-2-ylmethanol
Vzorec: C₈H₇NO₂
MW: 149,1467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368333
CAS číslo: 77186-95-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR