1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%
Dodavatel: Apollo Scientific
OR902251-1G
OR902251-250MG
APOSOR902251-1GEA
8010
CZK
APOSOR902251-1G
APOSOR902251-250MG
1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%
1-(Pyrrolidin-2-ylmethyl)pyrrolidine
Vzorec:
C₉H₁₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD06653330 CAS číslo: 195311-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...