Tisk…

Methyl isoquinoline-4-carboxylate 95%

Dodavatel: Apollo Scientific

OR902212-1G OR902212-250MG
APOSOR902212-1GEA 2570 CZK
APOSOR902212-1G APOSOR902212-250MG
Methyl isoquinoline-4-carboxylate 95%
Methyl isoquinoline-4-carboxylate
Vzorec: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD11111833
CAS číslo: 20317-40-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR