N-Methyl-4-(methylamino)picolinamide 95%
Dodavatel: Apollo Scientific
OR902039-1G
OR902039-25G
OR902039-5G
APOSOR902039-1GEA
1490
CZK
APOSOR902039-1G
APOSOR902039-25G
APOSOR902039-5G
N-Methyl-4-(methylamino)picolinamide 95%
N-Methyl-4-(methylamino)picolinamide
Vzorec:
C₈H₁₁N₃O Storage Temperature: Ambient |
MDL Number:
MFCD11504981 CAS číslo: 1065074-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...